Learn More
HuC/HuD Rabbit anti-Zebrafish, Polyclonal, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA5119545
Description
Store as concentrated solution. Centrifuge briefly prior to opening vial. For short-term storage (1-2 weeks), store at 4°C. For long-term storage, aliquot and store at -20°C or below. Avoid multiple freeze-thaw cycles.
HuC/HuD Polyclonal antibody specifically detects HuC/HuD in Zebrafish samples. It is validated for Immunohistochemistry - Free Floating, Immunohistochemistry (Paraffin), Western Blot
Specifications
HuC/HuD | |
Polyclonal | |
Unconjugated | |
ELAVL4 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
30732, 30737 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot | |
0.93 mg/mL | |
PBS with 20% glycerol and 0.025% Proclin 300; pH 7 | |
ELAV (embryonic lethal, abnormal vision, Drosophila)-like 3 (Hu antigen C); ELAV (embryonic lethal, abnormal vision, Drosophila)-like 4 (Hu antigen D); ELAV like neuron-specific RNA binding protein 3; ELAV like neuron-specific RNA binding protein 4; ELAV like RNA binding protein 4; elavC; ELAVL3; elavl3 protein; elavl4; ELAV-like protein 3; ELAV-like protein 4; elav-related C; elav-related D; elrc; elrd; embryonic lethal, abnormal vision-related C; embryonic lethal, abnormal vision-related D; etID309927.3; Hu antigen C; Hu antigen D; hu-antigen C; Hu-antigen D; HuC; HUCL; hud; id:ibd1248; Paraneoplastic cerebellar degeneration-associated antigen; paraneoplastic encephalomyelitis antigen HuD; Paraneoplastic limbic encephalitis antigen 21; PLE21; PNEM; protein elrD; ribonucleoprotein; RNA-binding protein HuD; wu:fb77b03; wu:fc24h01; zHuC | |
Recombinant protein encompassing a sequence within the center region of zebrafish Elavl3+4. The exact sequence is proprietary. | |
200 μL | |
Primary | |
Zebrafish | |
Antibody | |
IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.