Learn More
Novus Biologicals™ Human Adipose Membrane Tissue Lysate (Adult Membrane Normal)
Brand:  Novus Biologicals™ NB820-60566
Product Code. 18202865
  • 6051.68 DKK / 0.10mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Human Adipose Membrane Tissue Lysate (Adult Membrane Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
0.1 mg
Membrane Protein - Human Adult Normal Tissue: Adipose
Normal
Human
Human Adipose Membrane Tissue Lysate (Adult Normal)
Adult
Adipose
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Membrane
Human
1.6 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only