missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human AHRR Partial ORF (NP_065782, 617 a.a. - 715 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00057491-Q01.25ug
This item is not returnable.
View return policy
Description
Dioxin is a teratogen that exerts its effects through the arylhydrocarbon receptor in conjunction with the receptor's binding partner, arylhydrocarbon receptor nuclear translocator. The protein encoded by this gene represses signal transduction by the arylhydrocarbon receptor by competing with the arylhydrocarbon receptor nuclear translocator for binding to the arylhydrocarbon receptor. Expression of the repressor is stimulated by the receptor/translocator heterodimer, thereby regulating receptor function through a negative feedback mechanism. In addition, the encoded protein can bind to nuclear factor kappa-B. [provided by RefSeq]
Sequence: RATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSALATLVPPQASGCTFLPSpecifications
NP_065782 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RATAGRSRELTPFHPAHCACLEPTDGLPQSEPPHQLCARGRGEQSCTCRAAEAAPVVKREPLDSPQWATHSQGMVPGMLPKSALATLVPPQASGCTFLP | |
RUO | |
AHRR | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57491 | |
AHRR (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
AHH/AHHR/KIAA1234/MGC167813/MGC176630/bHLHe77 | |
AHRR | |
Recombinant | |
wheat germ expression system |