Learn More
Invitrogen™ Human BCAR3 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP1467
Description
BCAR3 Synthetic Peptide, PEP-1467, from Invitrogen.
Breast tumors are initially dependent on estrogens for growth and progression and can be inhibited by anti-estrogens such as tamoxifen. However, breast cancers progress to become anti-estrogen resistant. Breast cancer anti-estrogen resistance gene 3 was identified in the search for genes involved in the development of estrogen resistance. The gene encodes a component of intracellular signal transduction that causes estrogen-independent proliferation in human breast cancer cells. The protein contains a putative src homology 2 (SH2) domain, a hall mark of cellular tyrosine kinase signaling molecules, and is partly homologous to the cell division cycle protein CDC48.
Specifications
8412 | |
purified | |
Human BCAR3 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O75815 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BCAR3 | |
0.02% sodium azide | |
50 μg | |
PBS |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.