missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human Bcl-10 Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP0056
This item is not returnable.
View return policy
Description
BCL-10 Synthetic Peptide, PEP-0056, from Invitrogen.
BCL-10 is an N-terminal CARD (Caspase Recruitment Domain) containing protein that is involved in the adaptive immune response. It promotes apoptosis, pro-caspase-9 maturation, and activation of NF-kappa-B via NIK and IKK. BCL-10 is also an adapter protein between upstream TNFR1-TRADD-RIP complex and the downstream NIK-IKK-IKAP complex. It is a substrate for MALT1. Mutations in the gene can lead to Immmunodeficiency 37 or Lymphoma, mucosa-associated lymphoid type.
Specifications
8915 | |
purified | |
Human Bcl-10 | |
-20°C | |
Liquid | |
Blocking Assay, Control | |
No | |
Peptide | |
RUO - research use only |
Human | |
O95999 | |
200 μg/mL | |
Synthetic Peptide | |
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2 | |
BCL10 | |
0.02% sodium azide | |
50 μg | |
PBS |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Invitrogen™ Human Bcl-10 Synthetic Peptide > 50 μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction