missing translation for 'onlineSavingsMsg'
Learn More

Human C-Reactive Protein/CRP Antibody, R&D Systems™
GREENER_CHOICE

Mouse Monoclonal Antibody

1405.00 DKK - 3180.00 DKK

Specifications

Antigen C-ReactiveProtein/CRP
Clone 987312
Dilution ELISA
Applications ELISA
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18746538
GREENER_CHOICE
View Documents
R&D Systems
MAB17072-SP
25 ÎĽg
1405.00 DKK
25µg
Please to purchase this item. Need a web account? Register with us today!
18722328
GREENER_CHOICE
View Documents
R&D Systems
MAB17072-100
100 ÎĽg
3180.00 DKK
100µg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

C-Reactive Protein/CRP Monoclonal specifically detects C-Reactive Protein/CRP in Human samples. It is validated for ELISA.
Specifications
Show More
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Human C-Reactive Protein/CRP Antibody, R&D Systems™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.