Learn More
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA is a secreted protein and is one of at least 9 paralogs of Acrp30/Adiponectin.
Brand:  R&D Systems™ RDC2241
Product Code. 18755968
  • 2460.00 DKK / 10µg
Explore more special offers
Description
Description
The Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA clone contains the complete ORF for C1qTNF4/CTRP4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The C1qTNF4/CTRP4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
hC1QTNF4
10 μg at 0.2 μg/μL
1003bp
114900
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_114115
cDNA is provided in 10 mM Tris-Cl
8.5
C1q and tumor necrosis factor related protein 4, C1qTNF4, complement C1q tumor necrosis factor-related protein 4, complement-c1q tumor necrosis factor-related protein 4, CTRP4ZACRP4, ZACRP4
Human
Product Title
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA > Host Species: Human