Learn More
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA is a secreted protein and is one of at least 9 paralogs of Acrp30/Adiponectin.
Brand: R&D Systems™ RDC2241
This item is not returnable.
View return policy
Description
The Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA clone contains the complete ORF for C1qTNF4/CTRP4, along with a Kozak consensus sequence for optimal translation initiation. The gene insert is flanked with convenient multiple cloning sites which can be used to easily cut and transfer the gene cassette into your desired expression vector. The C1qTNF4/CTRP4 insert is sequence verified and the entire plasmid DNA and insert translated sequences are provided.
Specifications
hC1QTNF4 | |
10 μg at 0.2 μg/μL | |
1003bp | |
114900 | |
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
10 μg |
NP_114115 | |
cDNA is provided in 10 mM Tris-Cl | |
8.5 | |
C1q and tumor necrosis factor related protein 4, C1qTNF4, complement C1q tumor necrosis factor-related protein 4, complement-c1q tumor necrosis factor-related protein 4, CTRP4ZACRP4, ZACRP4 | |
Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
R&D Systems™ Human CTRP4/C1qTNF4 (NP_114115) VersaClone cDNA > Host Species: Human
Spot an opportunity for improvement?Share a Content Correction