Learn More
R&D Systems™ Human DMBT1 (Unique) VersaClone cDNA
Contains gene inserts that are fully sequenced and produced with high quality
Brand:  R&D Systems™ RDC3108
Product Code. 18700967
  • 8810.00 DKK / 10µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
DMBT1/GP340
10 μg at 0.2 μg/μL
7273 bp
CRP-ductin, deleted in malignant brain tumors 1, deleted in malignant brain tumors 1 protein, Ebnerin, FLJ61058, Glycoprotein 340, gp-340, GP340, GP340muclin, Hensin, MGC164738, Muclin, SAG, Salivary agglutinin, Surfactant pulmonary-associated D-binding protein, Vomeroglandin
Human
unique
cDNA is provided in 10 mM Tris-Cl, pH 8.5
1755
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg