missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GDAP1 Full-length ORF (NP_001035808.1, 1 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Specifications
Accession Number | NP_001035808.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 54332 |
Molecular Weight (g/mol) | 59.9kDa |
Description
This gene encodes a member of the ganglioside-induced differentiation-associated protein family, which may play a role in a signal transduction pathway during neuronal development. Mutations in this gene have been associated with various forms of Charcot-Marie-Tooth Disease and neuropathy. Two transcript variants encoding different isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MRLNSTGEVPVLIHGENIICEATQIIDYLEQTFLDERTPRLMPDKESMYYPRVQHYRELLDSLPMDAYTHGCILHPELTVDSMIPAYATTRIRSQIGNTESELKKLAEENPDLQEAYIAKQKRLKSKLLDHDNVKYLKKILDELEKVLDQVETELQRRNEETPEEGQQPWLCGESFTLADVSLAVTLHRLKFLGFARRNWGNGKRPNLETYYERVLKRKTFNKVLGHVNNILISAVLPTAFRVAKKRAPKVLGTTLVVGLLAGVGYFAFMLFRKRLGSMILAFRPRPNYFSpecifications
NP_001035808.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
59.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MRLNSTGEVPVLIHGENIICEATQIIDYLEQTFLDERTPRLMPDKESMYYPRVQHYRELLDSLPMDAYTHGCILHPELTVDSMIPAYATTRIRSQIGNTESELKKLAEENPDLQEAYIAKQKRLKSKLLDHDNVKYLKKILDELEKVLDQVETELQRRNEETPEEGQQPWLCGESFTLADVSLAVTLHRLKFLGFARRNWGNGKRPNLETYYERVLKRKTFNKVLGHVNNILISAVLPTAFRVAKKRAPKVLGTTLVVGLLAGVGYFAFMLFRKRLGSMILAFRPRPNYF | |
RUO | |
GDAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
54332 | |
GDAP1 (Human) Recombinant Protein (P02) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
GDAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |