Learn More
Invitrogen™ eBioscience™ Human IL-1 beta Instant ELISA Kit, for in vitro diagnostic use
For the quantitative detection of human IL-1beta.
Brand:  Invitrogen™ BMS224INSTCE
Product Code. 15511757
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Includes:
  • Aluminum pouch(es) with a Microwell Plate coated with Monoclonal Antibody (murine) to human IL-1beta, Biotin-Conjugate (anti-IL-1beta monoclonal antibody), Streptavidin-HRP and Sample Diluent, lyophilized
  • Aluminum pouch(es) with a human IL-1beta Standard curve (coloured)
  • Wash Buffer Concentrate 20x (phosphate-buffered saline with 1% Tween 20)
  • Sample Diluent (Use when an external predilution of the samples is needed)
  • Substrate Solution (tetramethyl-benzidine)
  • Stop Solution (1M Phosphoric acid)
  • Control high, lyophilized
  • Control low, lyophilized
  • Adhesive Films
Description
Description
Pre-coated plate(s)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
0.7pg/mL
Cell Culture Supernatant, Plasma (Citrate, Heparin), Serum
ELISA
50 ÎĽL
IL-1 beta
Biotin
Human
128 Tests
Catabolin, Interleukin-1 beta, IL1F2
Certificates
Promotions
Promotions
Product Content Correction
Product Title
Invitrogen™ eBioscience™ Human IL-1 beta Instant ELISA Kit, for in vitro diagnostic use > Human IL-1 beta Instant ELISA Kit, for in vitro diagnostic use
For Research Use Only