missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IL-17E/IL-25 Antibody, R&D Systems™
Mouse Monoclonal Antibody
1725.00 DKK - 3380.00 DKK
Specifications
Antigen | IL-17E/IL-25 |
---|---|
Clone | 878210R |
Dilution | ELISA |
Applications | ELISA |
Classification | Monoclonal |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18119780
|
R&D Systems
MAB12581-SP |
25 ÎĽg |
1725.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18159580
|
R&D Systems
MAB12581-100 |
100 ÎĽg |
3380.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IL-17E/IL-25 Monoclonal specifically detects IL-17E/IL-25 in Human samples. It is validated for ELISA.Specifications
IL-17E/IL-25 | |
ELISA | |
Monoclonal | |
Supernatant | |
Human | |
Q9H293 | |
64806 | |
IgG2a | |
Protein A or G purified from cell culture supernatant | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
878210R | |
ELISA | |
Unconjugated | |
Mouse | |
Lyophilized from a 0.2ÎĽm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2ÎĽm filtered solution in PBS. | |
IL17E, IL-17E, IL25, IL-25, interleukin 25, Interleukin-17E, interleukin-25 | |
E. coli-derived recombinant human IL-17E Tyr33-Gly177 Accession # Q9H293 | |
Primary | |
Reconstitute at 0.5mg/mL in sterile PBS. | |
Detects human IL-17E in direct ELISAs. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human IL-17E/IL-25 Antibody, R&D Systems™