Learn More
Invitrogen™ Human IL-34 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP0714
Product Code. 15368920
  • 1006.00 DKK / 50µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
IL-34 Synthetic Peptide, PEP-0714, from Invitrogen.
This peptide corresponds to 13 amino acids near the center of human IL-34PEP-0714 can be used as a blocking peptide with polyclonal antibody PA5-20594.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
146433
purified
Human IL-34
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q6ZMJ4
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
IL34
0.02% sodium azide
50 μg
PBS