missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human IPP Partial ORF (NP_005888.1, 105 a.a. - 204 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003652-Q01.25ug
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the kelch family of proteins, which is characterized by a 50 amino acid repeat which interacts with actin. Transcript variants have been described but their full-length nature has not been determined. [provided by RefSeq]
Sequence: NNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEFSENYIHVHFLEVHSGEEFLALTKDQLIKILRSEELSIEDEYQVFLAASpecifications
NP_005888.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
NNVQELIIAADMLQLTEVVHLCCEFLKGQIDPLNCIGIFQFSEQIACHDLLEFSENYIHVHFLEVHSGEEFLALTKDQLIKILRSEELSIEDEYQVFLAA | |
RUO | |
IPP | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3652 | |
IPP (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KLHL27 | |
IPP | |
Recombinant | |
wheat germ expression system |