Learn More
R&D Systems™ Human Lysyl Oxidase/LOX (NP_002308.2) VersaClone cDNA
Brand:  R&D Systems™ RDC3248
Product Code. 18752617
  • 2285.00 DKK / 10µg
Specifications
LOX
4015
Store the unopened product at -20°C to -70°C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date.
10 μg
NP_002308.2
AAT10, lysyl oxidaseEC 1.4.3.13, MGC105112, protein-lysine 6-oxidase
Human
Certificates
Promotions
Promotions
Product Content Correction
Product Title
R&D Systems™ Human Lysyl Oxidase/LOX (NP_002308.2) VersaClone cDNA > 10 μg