missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat STAT6 aa 627-838 Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 1 publication
Brand: R&D Systems AF2167
This item is not returnable.
View return policy
Description
STAT6 Polyclonal specifically detects STAT6 in Human, Mouse, Rat samples. It is validated for Western Blot, Intracellular Staining by Flow Cytometry, CyTOF-ready.
Specifications
STAT6 | |
Polyclonal | |
Unconjugated | |
D12S1644, EC 2.4.1.227, EC 2.7.7.6, IL-4 Stat, IL-4-STAT, signal transducer and activator of transcription 6, signal transducer and activator of transcription 6, interleukin-4 induced, STAT, interleukin-4 induced, STAT, interleukin4-induced, STAT6B, STAT6C, transcription factor IL-4 STAT | |
Goat | |
Affinity Purified | |
RUO | |
6778 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Western Blot, Flow Cytometry, CyTOF | |
LYOPH | |
Western Blot 0.5 ug/mL, Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells, CyTOF-ready | |
STAT6 | |
E. coli-derived recombinant mouse STAT6 | |
100 μg | |
Primary | |
Detects human, mouse, and rat STAT6 in Western blots. | |
Human, Mouse, Rat | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human/Mouse/Rat STAT6 aa 627-838 Antibody, R&D Systems™ > 100μg; Unlabeled
Spot an opportunity for improvement?Share a Content Correction