missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Osterix/Sp7 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems MAB7547-SP
This item is not returnable.
View return policy
Description
Osterix/Sp7 Monoclonal specifically detects Osterix/Sp7 in Human samples. It is validated for Western Blot, Immunocytochemistry.
Specifications
Osterix/Sp7 | |
Monoclonal | |
LYOPH | |
Western Blot 1 ug/mL, Immunocytochemistry 8-25 ug/mL | |
Q8TDD2 | |
SP7 | |
E. coli-derived recombinant human Osterix/Sp7 Met19-Leu288 Accession # Q8TDD2 | |
25 μg | |
Primary | |
Detects human Osterix/Sp7 in direct ELISAs. | |
Human | |
Purified |
Western Blot, Immunocytochemistry | |
764704 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
MGC126598, OSX, OSXosterix, Sp7, Sp7 transcription factor, transcription factor Sp7, Zinc finger protein osterix | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
121340 | |
Sterile PBS to a final concentration of 0.5 mg/mL. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Human Osterix/Sp7 Antibody, R&D Systems™ > 25μg; Unlabeled
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction