Learn More
Novus Biologicals™ Human Ovary Tissue MicroArray (Tumor)
Tissue arrays are 4 - 6 um in thickness, and mounted on positively charged glass slides
Brand:  Novus Biologicals™ NBP2-42068
Product Code. 18602957
  • 5265.00 DKK / Pack of 5
Explore more special offers
This item is not returnable. View return policy
Description
Description
Specifications
Specifications
5 Slides
Human
Immunohistochemistry, Immunohistochemistry-Paraffin