missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PI4KII Partial ORF (NP_060895.1, 383 a.a. - 477 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00055361-Q01.25ug
This item is not returnable.
View return policy
Description
Phosphatidylinositolpolyphosphates (PtdInsPs) are centrally involved in many biologic processes, ranging from cell growth and organization of the actin cytoskeleton to endo- and exocytosis. PI4KII phosphorylates PtdIns at the D-4 position, an essential step in the biosynthesis of PtdInsPs (Barylko et al., 2001 [PubMed 11244087]).[supplied by OMIM]
Sequence: LILPKISDPNFVKDLEEDLYELFKKDPGFDRGQFHKQIAVMRGQILNLTQALKDNKSPLHLVQMPPVIVETARSHQRSSSESYTQSFQSRKPFFSSpecifications
NP_060895.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.19kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LILPKISDPNFVKDLEEDLYELFKKDPGFDRGQFHKQIAVMRGQILNLTQALKDNKSPLHLVQMPPVIVETARSHQRSSSESYTQSFQSRKPFFS | |
RUO | |
PI4K2A | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
55361 | |
PI4KII (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp761G1923/PI4KII/PIK42A/RP11-548K23.6 | |
PI4K2A | |
Recombinant | |
wheat germ expression system |