missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STEMCELL Technologies Human Recombinant bFGF
Basic fibroblast growth factor

1139.40 DKK - 10563.75 DKK
Specifications
Name | Basic fibroblast growth factor, FGF-β, FGF2, Fibroblast growth factor-basic, HBGF-2 |
---|---|
Cell Type | Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells |
Purity or Quality Grade | >95% |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15614959
![]() |
STEMCELL Technologies
78003 |
50 ÎĽg |
2805.30 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16883713
![]() |
STEMCELL Technologies
78003.2 |
1000 ÎĽg |
10563.75 DKK
1000µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17119321
![]() |
STEMCELL Technologies
78003.1 |
10 ÎĽg |
1139.40 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
- Basic fibroblast growth factor (bFGF) is a prototypic member of the fibroblast growth factor family
- Cytokines in the FGF family possess broad mitogenic and cell survival activities (Folkman & Klagsbrun; Kimelman & Kirschner) and are involved in a variety of biological processes including cell proliferation, differentiation, survival, and apoptosis (Folkman & Klagsbrun; Klagsbrun; Rifkin & Moscatelli)
- bFGF has the β-trefoil structure (Ponting & Russell), binds to the four FGF receptor (FGFR) family members, and activates JAK/STAT, PI3K, ERK1/2, and other receptor tyrosine kinase (RTK) signaling pathways
- It supports the maintenance of undifferentiated human embryonic stem cells (Xu et al.; Kang et al.), stimulates human embryonic stem cells to form neural rosettes (Zhang et al.), and improves proliferation of human mesenchymal stem cells and enhances chondrogenic differentiation (Solchaga et al.)
Specifications
Basic fibroblast growth factor, FGF-β, FGF2, Fibroblast growth factor-basic, HBGF-2 | |
Brain Tumor Stem Cells; Endoderm, PSC-Derived; Hematopoietic Stem and Progenitor Cells; Mesenchymal Stem and Progenitor Cells; Mesoderm, PSC-Derived; Neural Cells, PSC-Derived; Neural Stem and Progenitor Cells; Neurons; Pluripotent Stem Cells; Prostate Cells | |
>95% |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
STEMCELL Technologies Human Recombinant bFGF