Learn More
Invitrogen™ Human S100A7L2 (aa 50-74) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (36%), Rat (36%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-63496 (PA5-63496. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications
Specifications
Accession Number | Q5SY68 |
Concentration | ≥5.0 mg/mL |
For Use With (Application) | Blocking Assay, Control |
Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
Gene ID (Entrez) | 645922 |
Name | Human S100A7L2 (aa 50-74) Control Fragment |
Quantity | 100 μL |
Regulatory Status | RUO |
Gene Alias | protein S100-A7-like 2; S100 calcium binding protein A7 like 2; S100 calcium binding protein A7-like 2; S100 calcium-binding protein A7-like 2; S100a7b; S100A7L2 |
Common Name | S100A7L2 |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.