missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SELENBP1 Full-length ORF (NP_003935.2, 1 a.a. - 472 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00008991-P01.25ug
This item is not returnable.
View return policy
Description
This gene product belongs to the selenium-binding protein family. Selenium is an essential nutrient that exhibits potent anticarcinogenic properties, and deficiency of selenium may cause certain neurologic diseases. It has been proposed that the effects of selenium in preventing cancer and neurologic diseases may be mediated by selenium-binding proteins. The exact function of this gene is not known. [provided by RefSeq]
Sequence: MATKCGNCGPGYSTPLEAMKGPREEIVYLPCIYRNTGTEAPDYLATVDVDPKSPQYCQVIHRLPMPNLKDELHHSGWNTCSSCFGDSTKSRTKLVLPSLISSRIYVVDVGSEPRAPKLHKVIEPKDIHAKCELAFLHTSHCLASGEVMISSLGDVKGNGKGGFVLLDGETFEVKGTWERPGGAAPLGYDFWYQPRHNVMISTEWAAPNVLRDGFNPADVEAGLYGSHLYVWDWQRHEIVQTLSLKDGLIPLEIRFLHNPDAAQGFVGCALSSTIQRFYKNEGGTWSVEKVIQVPPKKVKGWLLPEMPGLITDILLSLDDRFLYFSNWLHGDLRQYDISDPQRPRLTGQLFLGGSIVKGGPVQVLEDEELKSQPEPLVVKGKRVAGGPQMIQLSLDGKRLYITTSLYSAWDKQFYPDLIREGSVMLQVDVDTVKGGLKLNPNFLVDFGKEPLGPALAHELRYPGGDCSSDIWISpecifications
NP_003935.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
78.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ13813/LPSB/SP56/hSBP/hSP56 | |
SELENBP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
8991 | |
SELENBP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MATKCGNCGPGYSTPLEAMKGPREEIVYLPCIYRNTGTEAPDYLATVDVDPKSPQYCQVIHRLPMPNLKDELHHSGWNTCSSCFGDSTKSRTKLVLPSLISSRIYVVDVGSEPRAPKLHKVIEPKDIHAKCELAFLHTSHCLASGEVMISSLGDVKGNGKGGFVLLDGETFEVKGTWERPGGAAPLGYDFWYQPRHNVMISTEWAAPNVLRDGFNPADVEAGLYGSHLYVWDWQRHEIVQTLSLKDGLIPLEIRFLHNPDAAQGFVGCALSSTIQRFYKNEGGTWSVEKVIQVPPKKVKGWLLPEMPGLITDILLSLDDRFLYFSNWLHGDLRQYDISDPQRPRLTGQLFLGGSIVKGGPVQVLEDEELKSQPEPLVVKGKRVAGGPQMIQLSLDGKRLYITTSLYSAWDKQFYPDLIREGSVMLQVDVDTVKGGLKLNPNFLVDFGKEPLGPALAHELRYPGGDCSSDIWI | |
RUO | |
SELENBP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |