missing translation for 'onlineSavingsMsg'
Learn More
ACROBiosystems Human SIRP gamma / CD172g Protein, Fc Tag Product Code.: 15897697

ACROBiosystems Human SIRP gamma / CD172g Protein, Fc Tag

Product Code. 15897697
100 μg, 100µg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 15897697

Brand: ACROBiosystems SIGH5253100UG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Signal-regulatory protein gamma (SIRPG or SIRP-gamma) is also known as CD172 antigen-like family member B (CD172g), Signal-regulatory protein beta-2 (SIRPB2), is a single-pass type I membrane protein. SIRPG is expressed on CD4+ T-cells, CD8+ T-cells, CD56-bright natural killer (NK) cells, CD20+ cells, and all activated NK cells. SIRPG interacts with CD47. On binding with CD47, CD172g mediates cell-cell adhesion.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 63.7 kDa. The protein migrates as 67-85 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.

Specifications

Accession Number AAH64532
Molecular Weight (g/mol) 63.7 kDa
Quantity 100 μg
Source Human SIRP gamma, Fc Tag (SIG-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 29 - Ser 364 (Accession # AAH64532).
Immunogen Glu 29 - Ser 364
Storage Requirements -20°C
Gene Alias SIRPG,CD172g,SIRPB2,SIRP-gamma,SIRP-b2,SIRP-beta-2,Cripto-1
Common Name SIRP gamma
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Buffer 50 mM Tris, 100 mM Glycine, pH7.5
Endotoxin Level Less than 1.0 EU / μg by the LAL method.
Expression System HEK293
Form Powder
Protein Subtype Immune Checkpoint Proteins
Purity or Quality Grade >95% as determined by SDS-PAGE
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ACROBiosystems Human SIRP gamma / CD172g Protein, Fc Tag

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.