Learn More
Invitrogen™ Human SNX1 (aa 139-155) Synthetic Peptide
Synthetic Peptide
Brand: Invitrogen™ PEP286
This item is not returnable.
View return policy
Description
Human SNX1 (residues 139-155) Synthetic Peptide, PEP-286, from Invitrogen.
PEP-286 is a 17 amino acid synthetic peptide whose sequence is derived from human SNX1 protein. The sequence of this peptide is (amino to carboxy terminus): Q (139)-E-D-Q-F-D-L-T-V-G-I-T-D-P-E-K-I (155)This peptide may be used for neutralization and control experiments with the polyclonal antibody that reacts with this product, SNX1, catalog # PA1-836. Using a solution of peptide of equal volume and concentration to the corresponding antibody will yield a large molar excess of peptide (∼70-fold) for competitive inhibition of antibody-protein binding reactions.Reconstitute with 0.1 ml of distilled water.Specifications
6642 | |
purified | |
Human SNX1 (aa 139-155) | |
-20°C. Avoid freeze-thaw cycles. | |
Lyophilized | |
Control | |
Yes | |
Peptide | |
RUO - research use only |
Human | |
Q13596 | |
0.5 mg/mL | |
Synthetic Peptide | |
peptide with no preservative | |
SNX1 | |
No preservative | |
50 μg | |
Q(139) - E - D - Q - F - D - L - T - V - G - I - T - D - P - E - K - I (155) |