missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human SYT13 Partial ORF (NP_065877.1, 36 a.a. - 135 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
2665.00 DKK - 4040.00 DKK
Specifications
Accession Number | NP_065877.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 57586 |
Molecular Weight (g/mol) | 36.74kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16179186
|
Abnova™
H00057586-Q01.25ug |
25 ug |
4040.00 DKK
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16169186
|
Abnova™
H00057586-Q01.10ug |
10 ug |
2665.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
SYT13 belongs to the large synaptotagmin protein family. All synaptotagmins show type I membrane topology, with an extracellular N terminus, a single transmembrane region, and a cytoplasmic C terminus containing tandem C2 domains. Major functions of synaptotagmins include vesicular traffic, exocytosis, and secretion.[supplied by OMIM]
Sequence: KKGLLPRDQDPDLEKAKPSLLGSAQQFNVKKSTEPVQPRALLKFPDIYGPRPAVTAPEVINYADYSLRSTEEPTAPASPQPPNDSRLKRQVTEELFILPQSpecifications
NP_065877.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA1427 | |
SYT13 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
57586 | |
SYT13 (Human) Recombinant Protein (Q01) | |
KKGLLPRDQDPDLEKAKPSLLGSAQQFNVKKSTEPVQPRALLKFPDIYGPRPAVTAPEVINYADYSLRSTEEPTAPASPQPPNDSRLKRQVTEELFILPQ | |
RUO | |
SYT13 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |