missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Gibco™ Human TGF-beta 3 Recombinant Protein, PeproTech®
E. coli Recombinant Protein
594.00 DKK - 38370.00 DKK
Specifications
Accession Number | P10600 |
---|---|
For Use With (Application) | ELISA, Functional Assay, Western Blot |
Formulation | protein with no preservative |
Gene ID (Entrez) | 7043 |
Molecular Weight (g/mol) | 25 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17840653
|
Gibco™
100-36E-100UG |
100 μg |
7690.00 DKK
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17880643
|
Gibco™
100-36E-10UG |
10 μg |
1444.00 DKK
10µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17860653
|
Gibco™
100-36E-1MG |
1 mg |
38370.00 DKK
1mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17879882
|
Gibco™
100-36E-250UG |
250 μg |
13480.00 DKK
250µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17831023
|
Gibco™
100-36E-2UG |
2 μg |
594.00 DKK
2µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17890263
|
Gibco™
100-36E-500UG |
500 μg |
23050.00 DKK
500µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17890443
|
Gibco™
100-36E-50UG |
50 μg |
4575.00 DKK
50µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.Specifications
P10600 | |
protein with no preservative | |
25 | |
E. coli | |
RUO | |
ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3 | |
Human | |
Recombinant | |
Lyophilized |
ELISA, Functional Assay, Western Blot | |
7043 | |
Human TGF-beta 3 | |
-20°C | |
<1 EU/μg | |
TGFB3 | |
Unconjugated | |
ALDTNYCFRN LEENCCVRPL YIDFRQDLGW KWVHEPKGYY ANFCSGPCPY LRSADTTHST VLGLYNTLNP EASASPCCVP QDLEPLTILY YVGRTPKVEQ LSNMVVKSCK CS | |
≥ 98% by SDS-PAGE gel and HPLC analyses. |