Learn More
Novus Biologicals™ Human Throat Whole Tissue Lysate (Adult Whole Normal)
Brand:  Novus Biologicals™ NB820-59268
Product Code. 18242273
  • 4065.00 DKK / 1mg
Explore more special offers
This item is not returnable. View return policy
Description
Description
The Human Throat Whole Tissue Lysate (Adult Whole Normal) is derived from human. It has been validated for the following applications: Western Blot, Immunoprecipitation.
Specifications
Specifications
Western Blot
1 mg
Total Protein - Human Adult Normal Tissue: Throat
Normal
Human
Human Throat Whole Tissue Lysate (Adult Normal)
Adult
Digestive/Waste
HEPES (pH 7.9), MgCl2, KCl, EDTA, Sucrose, Glycerol, Sodium deoxycholate, NP-40, and a cocktail of protease inhibitors
Whole
Human
5 mg/mL
-80°C. Avoid freeze-thaw cycles.
Native
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Special Offers
Special Offers
For Research Use Only