missing translation for 'onlineSavingsMsg'
Learn More
ACROBiosystems Human TLR4 Protein, Fc Tag Product Code.: 17256580

ACROBiosystems Human TLR4 Protein, Fc Tag

Product Code. p-7232345
100 μg, 100µg
Encompass_Preferred
Click to view available options
Quantity:
1 mg
100 μg
Unit Size:
100µg
1mg
This item is not returnable. View return policy

Product Code. 17256580

Brand: ACROBiosystems TL4H5253100ug

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Toll-like receptor 4 (TLR4) is a member of the toll-like receptor (TLR) family and highly conserved receptors that recognize conserved pathogen-associated molecular patterns (PAMPs). TLR4 is expressed on the cell surface on both hematopoietic and nonhematopoietic cells, including endothelial cells], cardiac myocytes, and cells of the central nervous system (CNS). TLR4 has been regarded as the sensing receptor for gram-negative lipopolysaccharide (LPS). Further research indicated that TLR4also binds endogenous molecules produced as a result of tissue injury. Thus, TLR4 represents a key receptor on which both infectious and noninfectious stimuli converge to induce a proinflammatory response.

This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a calculated MW of 95.6 kDa.

Specifications

Molecular Weight (g/mol) 95.6 kDa
Quantity 100 μg
Source Human TLR4, Fc Tag (TL4-H5253) is expressed from human 293 cells (HEK293). It contains AA Glu 24 - Lys 631 (Accession # O00206-1).
Immunogen Glu 24 - Lys 631
Storage Requirements -20°C
Gene Alias TLR4/hToll/CD284/Toll-like receptor 4
Common Name TLR4
Conjugate Unconjugated
Species Human
Protein Tag Fc Tag
Expression System HEK293
Form Powder
Protein Subtype Other Recombinant Proteins
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title
ACROBiosystems Human TLR4 Protein, Fc Tag

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.