Learn More
Invitrogen™ Human TMEM70 Synthetic Peptide
Synthetic Peptide
Brand:  Invitrogen™ PEP1029
Product Code. 15265029
  • 1004.00 DKK / 50µg
Explore more special offers
This item is not returnable. View return policy
Description
Description
TMEM70 Synthetic Peptide, PEP-1029, from Invitrogen.
This peptide corresponds to 14 amino acids near the center of human TMEM70.PEP-1029 can be used as a blocking peptide with polyclonal antibody PA5-20915.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
54968
purified
Human TMEM70
-20°C
Liquid
Blocking Assay, Control
No
Peptide
RUO - research use only
Human
Q9BUB7
200 μg/mL
Synthetic Peptide
PBS with 0.1% BSA and 0.02% sodium azide; pH 7.2
TMEM70
0.02% sodium azide
50 μg
PBS