Learn More
Invitrogen™ Human UBA52 (aa 74-128) Control Fragment Recombinant Protein
Description
Highest antigen sequence indentity to the following orthologs: Mouse (100%), Rat (100%). This recombinant protein control fragment may be used for blocking experiments with the corresponding antibody, PA5-66029 (PA5-66029. In IHC/ICC and WB experiments, we recommend a 100x molar excess of the protein fragment control based on the concentration and the molecular weight. Pre-incubate the antibody-protein control fragment mixture for 30 min at room temperature.

Specifications
Specifications
Accession Number | P62987 |
Concentration | ≥5.0 mg/mL |
For Use With (Application) | Blocking Assay, Control |
Formulation | 1 M urea, PBS with no preservative; pH 7.4 |
Gene ID (Entrez) | 7311 |
Name | Human UBA52 (aa 74-128) Control Fragment |
Quantity | 100 μL |
Regulatory Status | RUO |
Gene Alias | 60 S ribosomal protein L40; CEP52; D8Ertd21e; Gm1863; HUB L40; HUBCEP52; L40; Large ribosomal subunit protein eL40; MGC127041; RPL40; RPS27A; Uba52; UBB; Ubc; Ubcep2; Ubiquitin; Ubiquitin A-52 residue ribosomal protein fusion product 1; ubiquitin and ribosomal protein L40; ubiquitin carboxyl extension protein 52; ubiquitin/60 S ribosomal fusion protein; ubiquitin-52 amino acid fusion protein; ubiquitin-60 S ribosomal protein L40; ubiquitin-CEP52; Unknown (protein for MGC:127041) |
Common Name | UBA52 |
Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.