Learn More
Proteintech™ HumanKine™ Recombinant Human Wnt3A Protein
Shop All Proteintech Products
Click to view available options
Quantity:
10 μg
100 μg
5 μg
Unit Size:
100µg
10µg
5µg
Description
- Results in authentic human proteins with native glycosylation, folding and increased stability, activity and purity
Specifications
Specifications
Gene ID (Entrez) | 89780 |
Molecular Weight (g/mol) | 38 to 42 kDa |
Quantity | 100 μg |
Species | Human |
Reconstitution | Briefly centrifuge the vial before opening. It is recommended to reconstitute the protein to ≤ 100 μg/mL in sterile 1 x PBS containing 0.1% endotoxin-free recombinant human serum albumin (HSA). |
Content And Storage | Lyophilized proteins are stable for 1 year from the date of receipt if stored between -20 to -80°C. Upon reconstitution, it is recommended the solution be stored at 4°C for short term or at -20 to -80°C for long term. Repeated freeze thaw cycles should be avoided with reconstituted products. |
Activity | Typically ≤ 20 ng/mL EC50 |
Expression System | Human embryonic kidney 293 (HEK293) human cell expression system |
Form | Lyophilized |
Purity or Quality Grade | > 90% |
Product Content Correction
Product Title
Proteintech™ HumanKine™ Recombinant Human Wnt3A Protein >
Spot an opportunity for improvement?Share a Content Correction