Learn More
Cytiva HyClone™ CDM4MAb Powdered Cell Culture Media
Click to view available options
Quantity:
5 L
Unit Size:
5 L
Description
HyClone CDM4MAb is a chemically defined and animal-derived component-free (ADCF) medium. This regulatory-friendly medium is designed to increase process yields for the manufacture of antibodies and antibody fragments for therapeutic use in a variety of engineered hybridoma and myeloma cell lines.
- Animal-derived component-free.
- Chemically defined formulation
- Designed for high cell yield and monoclonal antibody (MAb) production.
- Allows for direct or sequential adaptation
- Manufactured from traceable components according to cGMP guidelines
Specifications
Specifications
Form | Powder |
Sterility | Non-sterile |
Without Additives | No Glutamine, No Poloxamer 188 |
Quantity | 5 L |
Content And Storage | 2 to 8°C, away from light and moisture |
Product Content Correction
Product Title
Cytiva HyClone™ CDM4MAb Powdered Cell Culture Media >
Spot an opportunity for improvement?Share a Content Correction