Learn More
Cytiva HyClone™ Cell Boost™ 2 Supplement
Fed-batch medium supplement that contains amino acids, and glucose to improve productivity and cell growth
Brand: Cytiva SH31114.01
807.50 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), and Protein-Free
- Designed to support modern cell culture manufacturing processes
- Supplements cell culture with amino acids and glucose
- Does not contain L-glutamine, poloxamer 188, or hydrolysates
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
Clear | |
Liquid | |
≤ 1 EU/mL | |
Improving Productivity and Cell Growth in Fed-Batch Cultures | |
9.9 to 10.4 | |
500 mL |
CHO, PER.C6&trade, | |
Cell Boost Supplement | |
Bottle | |
300 to 367 mOsmol/kg | |
HyClone™ |