Learn More
Cytiva HyClone™ Cell Boost™ 4 Supplement
Fed-batch media supplement containing amino acids, vitamins, glucose, trace elements, and growth factors for CHO cells
Brand: Cytiva SH31116.01
912.00 DKK valid until 2025-01-31
Use promo code "23894" to get your promotional price.
This item is not returnable.
View return policy
Description
- Chemically Defined (CD), Animal-Derived Component-Free (ADCF), Protein-Free, and Hydrolysate-Free
- Supports fed-batch cultures with amino acids, vitamins, glucose, trace elements, and growth factors
- Developed for recombinant protein production with various cell lines including Chinese Hamster Ovary (CHO) cell line
- Designed to support modern cell culture manufacturing processes in research and bioprocessing applications
- Manufactured to meet cGMP manufacturing standards and QC specifications
- Available as a ready-to-use liquid feed solution
Specifications
CHO | |
Cell Boost Supplement | |
Improving Productivity and Cell Yield in Fed-Batch Cultures | |
500 mL |
Liquid | |
Bottle | |
HyClone™ |