missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytiva HyClone™ Iron-Supplemented Newborn Calf Serum, New Zealand Origin
Contains three to four times as much available iron and transferrin as FBS or equine serum
Brand: Cytiva SH30626.02IR30-40
This item is not returnable.
View return policy
Description
- Carefully collected, processed, and filtered in New Zealand to ensure safety against bovine diseases
- Contains three to four times as much available iron and transferrin as FBS or equine serums
- Collected from cattle that are typically less than 10 days old
Specifications
<10 days | |
Newborn Calf Bovine Serum | |
Sterile | |
Irradiated (30-40 kGy) | |
HyClone™ |
New Zealand | |
500 mL | |
Sequential Triple-filtered, 100 nm | |
Yes |