missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid, 0.5M, Chem-Lab Product Code.: 10010472
compliance-icons

Hydrochloric Acid, 0.5M, Chem-Lab

Product Code. 10010472
5 L, PE
Click to view available options
Quantity:
5 L
10 L
Packaging:
PE
This item is not returnable. View return policy

Product Code. 10010472

Brand: Chem Lab CL05.0310.5000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

For laboratory use, Titrations, ASTM D-94, D-128

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acidShow MoreShow Less
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

Concentration 18.23g HCl / 1L H2O
Density 1.01g/mL
Packaging PE
Normality 0.5N (±0.001 at 20°C)
Quantity 5 L
UN Number UN1789
Formula Weight 36.46g/mol
Chemical Name or Material Hydrochloric acid
compliance-icons
Product Identifier
  • Hydrochloric acid 0.5 mol/l
Signal Word
  • Warning
Hazard Category
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H290-May be corrosive to metals.
Precautionary Statement
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : HCl/H2O
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid, 0.5M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.