Learn More
Hydrochloric acid, 6.0N Standardized Solution, Thermo Scientific Chemicals
Description
6.0N HClsolution acts as a standard solution in volumetric titrations. In addition, it has industrial applications such as chemical transformations in hydrochlorination of rubber and production of vinyl and alkyl halide. In the semiconductor industry, it is used to etch semiconductor crystal and to purify silicon.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Quantity | 20 L |
Vapor Pressure | 23 hPa (17mm Hg) at 20°C |
Physical Form | Liquid |
Color | Colorless to Yellow |
Solubility | Miscible with water, methanol,ethanol and ether |
Formula Weight | 36.46 |
Concentration or Composition (by Analyte or Components) | Hydrochloric acid: 49.6%; Water: 50.4% |
Chemical Name or Material | Hydrochloric acid |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.