Learn More
Hydrochloric Acid Solution, 0.1M, Fisher Chemical™ Product Code.: 12970754
Hydrochloric Acid Solution, 0.1M, Fisher Chemical™
BIOPHARMA
Product Code. p-8014546 Shop All Fisher Chemical Products
100 mL, 100mL
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
Product Code. 12970754
Brand: Fisher Chemical SOL 113 100
All Our Certified Reference Materials and Reagents are produced under an ISO 9001 Quality Management System, also Accredited according to ISO/IEC 17025 and Accredited according to ISO Guide 34.
Specifications
Concentration 0.1mol/L
Packaging 100mL
Quantity 100 mL
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number Certificate Type Date Product Code
Lot Number1094752 Certificate TypeCertificate of Analysis Date30/03/2025 Product CodeSOL 113 100
Product Content Correction
Product Title
Hydrochloric Acid Solution, 0.1M, Fisher Chemical™ >