missing translation for 'onlineSavingsMsg'
Learn More
Hydrochloric Acid Solution, 3M, Chem-Lab Product Code.: 11964044
compliance-icons compliance-icons

Hydrochloric Acid Solution, 3M, Chem-Lab

Product Code. 11964044
1 L, Polyethylene, 1 L
Click to view available options
Quantity:
1 L
Packaging:
Polyethylene
Unit Size:
1 L
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 11964044

Brand: Chem Lab CL05.0315.1000

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Used as a laboratory reagent and acidifying agent. Hydrochloric Acid Solution, 1M, Chem-Lab contains 109.383g HCl per liter of water. It is used for pH control, titrations, the production of organic and inorganic compounds, and in the regeneration of ion exchange resins.

Hydrochloric acid is a corrosive, strong mineral acid with many industrial uses. It is a colorless, highly pungent solution of hydrogen chloride (HCl) in water. It is frequently used in pH buffer solutions, in the regeneration of ion exchange resins and in the production of chlorides and the synthesis of organic products.

Specifications

Appearance Liquid
pH <1
Appearance of Solution Liquid
Color Colorless
CAS 7647-01-0, 7732-18-5
Composition 109.383g HCl per 1L H2O
Concentration 3M
Density 1.05g/mL
Odor Odourless
Packaging Polyethylene
Normality 3N (±0.006, 20°C)
Quantity 1 L
Molecular Formula ClH
Physical Form Liquid
MDL Number MFCD00011324 MFCD00792839
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
Solubility Information Soluble in water
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
ChEBI CHEBI:17883
Formula Weight 36.46g/mol
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrochloric acid 3 mol/l
Signal Word
  • Warning
Hazard Category
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Substances and mixtures corrosive to metals Category 1
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H290-May be corrosive to metals.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P390-Absorb spillage to prevent material damage.
Supplemental information
  • MIXTURE LIST-Contains : Hydrochloric acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Hydrochloric Acid Solution, 3M, Chem-Lab >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.