missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium Hydroxide Solution, 32% Weight, AR, Chem-Lab
Click to view available options
Quantity:
5 L
10 L
Unit Size:
10 L
5 L
Description
Sodium hydroxide is used in many industries, mostly as a strong chemical base in the manufacture of pulp and paper, textiles, drinking water, soaps and detergents and as a drain cleaner.
Specifications
Specifications
pH | >13 |
Color | Colorless |
Boiling Point | 100°C |
Concentration | 32% by weight |
Density | 1.349g/mL |
Solubility Information | Water Soluble |
Quantity | 5 L |
Formula Weight | 40.00g/mol |
Concentration or Composition (by Analyte or Components) | 432gNaOH per 1L H2O |
Odor | Odorless |
Show More |
Safety and Handling

Product Identifier
- Sodium hydroxide 32 weight % solution a.r.
- Danger
- Skin corrosion/irritation Category 1 A
- Substances and mixtures corrosive to metals Category 1
- H314-Causes severe skin burns and eye damage.
- H290-May be corrosive to metals.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P308+P311-IF exposed or concerned: Call a POISON CENTER/doctor/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- MIXTURE LIST-Contains : NaOH/H2O
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Sodium Hydroxide Solution, 32% Weight, AR, Chem-Lab >
Spot an opportunity for improvement?Share a Content Correction