Learn More
Fisherbrand™ Hydrometer Rack, 10-Place
Rack for gentle handling and strong protection for fragile glass hydrometers.
870.00 DKK
Specifications
Product Type | Hydrometer Rack |
---|---|
Includes | Drip Tray |
Autoclavable | Not Autoclavable |
Holds | Hydrometers (Long), Hydrometers (Short) |
Material | Polypropylene |
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Color | Price | Quantity & Availability | ||||||
30130292
![]() |
Fisherbrand™
121002F |
Gray |
870.00 DKK
Each |
Estimated Shipment: 15-05-2025
Log in to see stock. |
||||||
30130293
![]() |
Fisherbrand™
121003F |
Blue |
870.00 DKK
Each |
Estimated Shipment: 22-05-2025
Log in to see stock. |
||||||
Description
The Hydrometer Rack accommodates up to 10 hydrometers with diameters up to 31 mm, ensuring safe storage, efficient drying, and secure transport.
Secure Organization
- Teardrop-Shaped Slots hold hydrometers securely in place, support a range of diameters, and ensure easy insertion and removal for convenient handling
- 3-Tier Setup provides enhanced stability and protection of hydrometer body
Clean and Convenient
- Built-in Handles provide a secure grip for safe and convenient transport
- Removable Drip Tray simplifies cleaning and maintenance, improving sanitation and protecting your workspace
Robust Design
- Stable Wide Base ensures increased stability and secure placement for hydrometers
- Durable Polypropylene Construction is easy to clean, long-lasting, and fully recyclable
Specifications
Hydrometer Rack | |
Not Autoclavable | |
Polypropylene | |
3 Tiers | |
29.5 cm | |
23.4 cm |
Drip Tray | |
Hydrometers (Long), Hydrometers (Short) | |
10 | |
29.5 x 17.50 x 23.40 cm | |
17.50 cm |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Fisherbrand™ Hydrometer Rack, 10-Place