missing translation for 'onlineSavingsMsg'
Learn More

Ansell HyFlex™ 48-102 Polyurethane-Coated Lightweight Nylon Gloves

Product Code. 16099721
Click to view available options
Size:
6
7
8
9
10
11
This item is not returnable. View return policy
This item is not returnable. View return policy

Gloves feature a thin liner and coating to ensure dexterity and flexibility. HyFlex™ 48-102 gloves are ideal for extreme tactility where precision is required.

  • Abrasion resistant for an extended wear life
  • Thin liner and coating offer dexterity and flexibility
  • Extreme tactility where precision is required
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance (Europe) EN 420:2003 + A1:2009, REACH Compliant
Coating Material Polyurethane
Color Gray
Hand Specific
Material Nylon
Size 9
EN388 Standard 4121A
For Use With (Application) General Handling, Packaging, and Machine Tool Operation
Glove Cuff Style Knit
Series 48-102
Yarn Gauge 13
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.