Learn More
Hygiena PRO-Clean™ Rapid Protein Residue Test
Accurately monitor the cleanliness of food equipment surfaces to help ensure food safety and product quality. Simply swab a surface, release the reagent and if food residue is present the reagent will turn purple. Quickly validates hygiene of surface, allowing immediate corrective action to be taken.
Brand:  Hygiena 14-037-778
Encompass
Product Code. 12789336
Description
Description
  • Color change provides semi-quantitative measure of surface cleanliness
  • The more contamination present, the quicker the color change to purple and the darker the color
  • PRO-Clean detects protein residues left on a surface after cleaning
  • Results in under 10 minutes
  • No instrumentation required - simple color-changing swab device
  • Detects 80μg protein after 1 minute
  • Detects 50μg protein after 5 minutes
  • Detects 20μg protein after 10 minutes
  • Results color chart printed on label for easy interpretation
  • Patented Snap-Valve™ technology and premoistened swab bud
  • Write-on swab tube label
  • Quick and easy way to monitor HACCP/SSOP program
  • Instant results allow for immediate corrective action
  • No technical training required
Meat processors, Food processors, Restaurants and food services, Grocery stores
Specifications
Specifications
2°C to 25°C
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Hygiena PRO-Clean™ Rapid Protein Residue Test