Learn More
HOPEN Hygiene PP Gown with Collar, Navy Blue Product Code.: 17256760
Add to basket
HOPEN Hygiene PP Gown with Collar, Navy Blue
Product Code. 17256760
Medium
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
Product Code. 17256760
Brand: HOPEN 40.702/02
Add to basket
Add to basket
Hygiene gown with zip and left inside pocket.
  • Hygiene PP Gown
  • Classic with collar
  • Elasticated wrists
  • With one interior pocket on the left side
  • With zip fastener
  • 40 g/m2
  • Navy Blue
Specifications
Size Medium
Material Polypropylene
Color Navy
Cuff Style Elastic
Includes Collar
  • Multiple Manufacturers
    Save up to 60% on gloves, apparel, and more
    Promo Code: 24131
    Expires2025-06-08
Product Content Correction
Product Title
HOPEN Hygiene PP Gown with Collar, Navy Blue >