Learn More
Sampling Systems™ Hygienic Butterfly Valve Kit
These hygienic butterfly valves are manufactured from 316L stainless steel and are ideal for many applications, including GMP use.
1975.00 DKK - 2030.00 DKK
Specifications
Product Type | Valve Kit |
---|---|
For Use With (Application) | Dispensing Liquid |
For Use With (Equipment) | Drum with Sloping Base |
Sterility | Non-sterile |
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | ||||||
17857817
|
Sampling Systems™
A136-01A |
Valve with EPDM Seals |
1975.00 DKK
Each |
Estimated Shipment: 03-07-2025
Log in to see stock. |
||||||
17912102
|
Sampling Systems™
A136-01S |
Valve with Silicone Seals |
2030.00 DKK
Each |
Estimated Shipment: 10-06-2025
Log in to see stock. |
||||||
Description
- Comes with a silicone or EPDM seal
- Both seals are suitable for food and pharmaceutical use and come with relevant certification
- Easy to operate: Simply turn the handle to open or close
- Opening the handle slightly will mean you have reduced product flow
- Valves are ideal for liquids
Specifications
Valve Kit | |
Dispensing Liquid | |
Drum with Sloping Base | |
Non-sterile |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Product Title
Sampling Systems™ Hygienic Butterfly Valve Kit