Learn More
Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Column Adapters
Order these cartridge adapters when using various cartridge sizes on the glass block vacuum manifold.
Brand: Thermo Scientific™ 60104-259
746.51 DKK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Description
Use the Thermo Scientific™ HyperSep™ Glass Block Vacuum Manifold Cartridge Adaptors for using various sized solid phase extraction (SPE) cartridges on the glass block vacuum manifold for sample preparation.
Specifications
Column Adaptors for 1 mL, 3 mL , 6 mL SPE Columns, 15 pk | |
Glass Block Vacuum Manifold, 1 mL, 3 mL , 6 mL SPE Columns | |
Column Adaptors |
SPE Sample Preparation | |
HyperSep |
Special Offers
-
Multiple Manufacturers
Contract Testing Lab
Use promotion code 24138 to get amazing savings on products for your analysisPromo Code: 24138Expires: 2025-04-30
Days
4Hours
21Minutes
21Seconds
41