Learn More
Thermo Scientific™ HyperSep™ Pressure Manifold Installation Kit
Obtain all the parts needed to set up your Thermo Scientific™ HyperSep™ Positive Pressure Manifold.
Brand: Thermo Scientific™ 60104-272
2932.23 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
3071.80 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Description
Use the Thermo Scientific™ HyperSep™ Positive Pressure Manifold Installation Kit for all the parts needed to set up your manifold. The HyperSep Positive Pressure Manifold holds up to 48 SPE cartridges. Samples and solvent, added to the SPE cartridge, are extracted under positive pressure for an even flow at each position. Extracts are collected into 13 or 16mm tubes.
Specifications
Installation Kit, 1 pk | |
Positive Pressure Manifold | |
Installation Kit |
SPE | |
HyperSep |