Learn More
Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE Cartridges.
Brand:  Thermo Scientific™ 60303-408
2666.92 DKK valid until 2025-12-31
Use promo code "25339" to get your promotional price.
Product Code. 10076154
  • 2680.25 DKK / Each
Estimated Shipment: 29-01-2025
to see stock.
Explore more special offers
Description
Achieve superior recoveries for acidic analytes with Thermo Scientific™ HyperSep™ Retain AX SPE 96-Well Plates and Individual Wells, packed with a high capacity, high purity, highly porous polystyrene DVB material partially modified with quaternary amine functional groups. Their use in SPE sample preparation can significantly enhance the robustness and reliability of many biological, pharmaceutical, forensic and environmental applications.
Widely used in forensic and clinical analysis of THC and its metabolites, Hypersep Retain SPE wells conform to industry standard configurations and allow efficient routine processing by vacuum, positive pressure or centrifugation.
  • High, consistent recoveries
  • Ideal for large samples
  • Fast and easy sample preparation and method development
  • Reproducible, reliable sample preparation in a traditional format
  • pH stable 0 to 14
  • Available in a wide range of chemistries and bed weights
For Use With: The Thermo Scientific™ HyperSep-96™ System
TRUSTED_SUSTAINABILITY
Specifications
Specifications
96-Well plate; Bed weight 60 mg
30 to 50 μm
HyperSep Retain AX
Analysis of THC and its Metabolites
HyperSep
96-well Plate