missing translation for 'onlineSavingsMsg'
Learn More
Learn More
IAH1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
1485.92 DKK - 3498.83 DKK
Specifications
Antigen | IAH1 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Purified |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18310764
|
Bio-Techne
NBP3-04579-20UL |
20 μL |
1485.92 DKK
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
18352175
|
Bio-Techne
NBP3-04579-100UL |
100 μL |
3498.83 DKK
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IAH1 Polyclonal specifically detects IAH1 in Human, Mouse, Rat samples. It is validated for Western Blot.Specifications
IAH1 | |
Polyclonal | |
Purified | |
RUO | |
PBS with 50% glycerol, pH7.3. | |
285148 | |
IgG | |
Affinity purified |
Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
EC 3.1, isoamyl acetate-hydrolyzing esterase 1 homolog, isoamyl acetate-hydrolyzing esterase 1 homolog (S. cerevisiae), MGC102860, S | |
Recombinant fusion protein containing a sequence corresponding to amino acids 1-248 of human IAH1 (NP_001034702.1). | |
Primary | |
Store at -20°C. Avoid freeze-thaw cycles. |