Learn More
IKA™ ICC Basic Circulator
GREENER_CHOICE
ICC basic circulator with an integrated carrying handle that allows for comfortable transport and safe storage.
Brand:  IKA™ 0004134400
Product Code. 15449783
  • 11183.98 DKK / Each
Only null left
In Stock
Description
  • Temperature range: RT to 150°C
  • Temperature stability: ± 0.02 K
  • Powerful, speed-controlled pressure/suction pump
  • Holding clamp for attaching the circulator to a bath is included
  • Convenient carrying handle und integrated brackets
  • Large easy to read LED display for pump speed and temperature
  • Push and turn buttons for intuitive menu navigation
  • Adjustable safety circuit
  • Visual and acoustic safety warnings
  • USB / RS 232 interface for PC control and documentation using labworldsoft™, as well as for online firmware updates
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Class designation acc. DIN 12876: III
Temperature control in water baths
230/115 V
ICC Basic, main cables, USB 2.0 cable (A–micro B), Screwdriver (use for safety circuit), user guide, warranty card
USB interface, RS 232 interface
3.75 kg
50/60 Hz
Temperature stability DIN 12876: 0.02 ±K
20 x 14.5 x 34 cm
LED
Basic
Power input: 2100 W; Heat output: 2000 W
Ambient +10°K at 1000 rpm to 150°C
Certificates
Promotions
Promotions
Product Content Correction
Product Title
IKA™ ICC Basic Circulator > Dimensions (L x W x H): 20 x 14.5 x 34 cm