Learn More
BINDER™ ICH Light Module
ICH-Q1B light module
Brand:  BINDER™ 8012-2441
 View more versions of this product
Product Code. 30238258
  • 30951.90 DKK / Each
Estimated Shipment: 07-05-2025
to see stock.
Explore more special offers
Description
Module consists of:
  • VIS cassette
  • UVA cassette
  • Independent power supply
Specifications
Specifications
KB PRO 260, KB PRO 470, KBF 260, KBF 470, KBF PRO 260, KBF PRO 470
Glass
Lighting
Product Title
BINDER™ ICH Light Module > For use with KB PRO 260, KB PRO 470, KBF 260, KBF 470, KBF PRO 260, KBF PRO 470